Assay Details
Gene Symbol: ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213172.1 | NP_998337.1 | 6 - 7 | 732 | 56 | |
GenBank mRNA | BC066368.1 | - | 6 - 7 | 732 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
