Assay Details
Gene Symbol: arglu1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213216.2 | NP_998381.2 | 1 - 2 | 491 | 82 | |
GenBank mRNA | BC044383.1 | - | 1 - 2 | 491 | 82 | |
BC062839.1 | - | 1 - 2 | 479 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
