Assay Details
Gene Symbol: arglu1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213216.2 | NP_998381.2 | 3 - 4 | 798 | 87 | |
GenBank mRNA | BC044383.1 | - | 3 - 4 | 798 | 87 | |
BC062839.1 | - | 3 - 4 | 786 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
