Assay Details
| Gene Symbol: cab39l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213501.1 | NP_998666.1 | 4 - 5 | 652 | 87 | |
| GenBank mRNA | BC044172.1 | - | 4 - 5 | 652 | 87 | |
| BC164384.1 | - | 3 - 4 | 433 | 87 | ||
More Information
Set Membership: |
|

cab39l1
Genomic Map