Assay Details
| Gene Symbol: cab39l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213501.1 | NP_998666.1 | 5 - 6 | 822 | 79 | |
| GenBank mRNA | BC044172.1 | - | 5 - 6 | 822 | 79 | |
| BC164384.1 | - | 4 - 5 | 603 | 79 | ||
More Information
Set Membership: |
|

cab39l1
Genomic Map