Assay Details
| Gene Symbol: acot11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037572.1 | NP_001032661.1 | 7 | 848 | 152 | |
| GenBank mRNA | BC110102.1 | - | 7 | 848 | 152 | |
More Information
Set Membership: |
|

acot11b
Genomic Map