Assay Details
| Gene Symbol: rgs5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199962.2 | NP_956256.1 | 5 | 2718 | 147 | |
| GenBank mRNA | BC054594.1 | - | 6 | 2452 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rgs5a| Gene Symbol: rgs5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199962.2 | NP_956256.1 | 5 | 2718 | 147 | |
| GenBank mRNA | BC054594.1 | - | 6 | 2452 | 147 | |
Set Membership: |
|