Assay Details
| Gene Symbol: fitm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020498.1 | NP_001018334.1 | 2 | 656 | 135 | |
| GenBank mRNA | BC094293.1 | - | 2 | 656 | 135 | |
| BC154267.1 | - | 2 | 633 | 135 | ||
More Information
Set Membership: |
|

fitm2
Genomic Map