Assay Details
| Gene Symbol: slc25a15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080638.1 | NP_001074107.1 | 7 | 1053 | 149 | |
| GenBank mRNA | BC129398.1 | - | 7 | 1053 | 149 | |
More Information
Set Membership: |
|

slc25a15a
Genomic Map