Assay Details
| Gene Symbol: rnls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002607.1 | NP_001002607.1 | 3 | 345 | 89 | |
| XM_005169454.4 | XP_005169511.1 | 3 | 384 | 89 | ||
| GenBank mRNA | BC075985.1 | - | 3 | 345 | 89 | |
More Information
Set Membership: |
|

rnls
Genomic Map