Assay Details
Gene Symbol: nkiras1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC076289.1 | - | 4 | 1851 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: nkiras1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC076289.1 | - | 4 | 1851 | 142 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |