Assay Details
| Gene Symbol: uqcrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037108.3 | NP_001032185.1 | 4 | 759 | 104 | |
| GenBank mRNA | BC107819.1 | - | 4 | 547 | 104 | |
| EH440299.1 | - | 4 | 608 | 104 | ||
| EH443764.1 | - | 4 | 574 | 104 | ||
More Information
Set Membership: |
|

uqcrh
Genomic Map