Assay Details
Gene Symbol: prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105698.1 | NP_001099168.1 | 3 | 745 | 149 | |
GenBank mRNA | BC151993.1 | - | 3 | 745 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
