Assay Details
| Gene Symbol: tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098391.1 | NP_001091861.1 | 6 | 1754 | 143 | |
| GenBank mRNA | BC139898.1 | - | 6 | 1754 | 143 | |
More Information
Set Membership: |
|

tmem17
Genomic Map