Assay Details
| Gene Symbol: prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076623.1 | NP_001070091.1 | 11 | 1251 | 144 | |
| XM_021471498.1 | XP_021327173.1 | 11 | 1280 | 144 | ||
| XM_021471499.1 | XP_021327174.1 | 11 | 1301 | 144 | ||
| GenBank mRNA | BC124401.1 | - | 11 | 1251 | 144 | |
| BC155668.1 | - | 11 | 1225 | 144 | ||
More Information
Set Membership: |
|

prkar1b
Genomic Map