Assay Details
| Gene Symbol: hmx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045371.1 | NP_001038836.2 | 2 | 654 | 99 | |
| GenBank mRNA | BC122454.1 | - | 2 | 654 | 99 | |
| EU050649.1 | - | 2 | 556 | 99 | ||
More Information
Set Membership: |
|

hmx4
Genomic Map