Assay Details
| Gene Symbol: ube2v1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037402.1 | NP_001032479.1 | 4 | 857 | 83 | |
| GenBank mRNA | BC107635.1 | - | 4 | 857 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ube2v1| Gene Symbol: ube2v1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037402.1 | NP_001032479.1 | 4 | 857 | 83 | |
| GenBank mRNA | BC107635.1 | - | 4 | 857 | 83 | |
Set Membership: |
|