Assay Details
Gene Symbol: nrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040326.1 | NP_001035416.1 | 17 | 3648 | 137 | |
XM_009297197.3 | XP_009295472.1 | 17 | 3906 | 137 | ||
XM_009297199.3 | XP_009295474.1 | 17 | 3886 | 137 | ||
GenBank mRNA | BC115224.1 | - | 17 | 3648 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
