Assay Details
Gene Symbol: hs2st1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080670.1 | NP_001074139.1 | 7 | 1552 | 97 | |
GenBank mRNA | BC129165.1 | - | 7 | 1552 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
