Assay Details
| Gene Symbol: tuba1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194388.2 | NP_919369.2 | 4 | 1462 | 161 | |
| GenBank mRNA | AF029250.1 | - | 3 | 1351 | 161 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tuba1b| Gene Symbol: tuba1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194388.2 | NP_919369.2 | 4 | 1462 | 161 | |
| GenBank mRNA | AF029250.1 | - | 3 | 1351 | 161 | |
Set Membership: |
|