Assay Details
Gene Symbol: ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213130.2 | NP_998295.2 | 5 | 964 | 159 | |
GenBank mRNA | BC053273.1 | - | 5 | 945 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
