Assay Details
| Gene Symbol: kctd15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003527.1 | 947 | 124 | |||
| GenBank mRNA | BC078294.1 | - | 947 | 124 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kctd15b| Gene Symbol: kctd15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003527.1 | 947 | 124 | |||
| GenBank mRNA | BC078294.1 | - | 947 | 124 | ||
Set Membership: |
|