Assay Details
| Gene Symbol: spry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003842.2 | NP_001003842.2 | 1 | 1052 | 143 | |
| XM_005160285.4 | XP_005160342.1 | 2 | 1597 | 143 | ||
| GenBank mRNA | BC095266.1 | - | 1 | 1052 | 143 | |
More Information
Set Membership: |
|

spry2
Genomic Map