Assay Details
| Gene Symbol: ubb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013272.2 | NP_001013290.2 | 1849 | 141 | ||
| GenBank mRNA | AY394942.1 | - | 4 | 893 | 141 | |
| BC069831.1 | - | 1849 | 141 | |||
| BC093444.1 | - | 1850 | 141 | |||
More Information
Set Membership: |
|

ubb
Genomic Map