Assay Details
| Gene Symbol: thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037566.1 | NP_001032655.1 | 8 | 1247 | 76 | |
| GenBank mRNA | BC110121.1 | - | 8 | 1247 | 76 | |
More Information
Set Membership: |
|

thnsl2
Genomic Map