Assay Details
| Gene Symbol: alg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104936.2 | NP_001098406.2 | 3 | 1801 | 97 | |
| XM_017351332.2 | XP_017206821.1 | 3 | 1851 | 97 | ||
| GenBank mRNA | BC151888.1 | - | 3 | 1750 | 97 | |
| BC152225.1 | - | 3 | 1767 | 97 | ||
More Information
Set Membership: |
|

alg2
Genomic Map