Assay Details
Gene Symbol: ahr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131264.1 | NP_571339.1 | 11 | 5649 | 131 | |
GenBank mRNA | AF063446.1 | - | 11 | 5649 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
