Assay Details
| Gene Symbol: twsg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153672.2 | NP_705958.1 | 846 | 83 | ||
| GenBank mRNA | BC103719.1 | - | 876 | 83 | ||
| BC118680.1 | - | 846 | 83 | |||
More Information
Set Membership: |
|

twsg1b
Genomic Map