Assay Details
Gene Symbol: sst1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183070.1 | NP_898893.1 | 2 | 252 | 68 | |
GenBank mRNA | AF435965.1 | - | 2 | 252 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
