Assay Details
Gene Symbol: twf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199725.1 | NP_956019.1 | 951 | 61 | ||
GenBank mRNA | BC058050.1 | - | 951 | 61 | ||
BC067628.1 | - | 935 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
