Assay Details
| Gene Symbol: jun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199987.1 | NP_956281.1 | 1 | 1554 | 71 | |
| GenBank mRNA | BC056597.1 | - | 1 | 1554 | 71 | |
| BC065976.1 | - | 1 | 1590 | 71 | ||
More Information
Set Membership: |
|

jun
Genomic Map