Assay Details
Gene Symbol: mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205640.1 | NP_991203.1 | 5 | 548 | 78 | |
GenBank mRNA | AY072811.1 | - | 5 | 548 | 78 | |
BC162742.1 | - | 5 | 518 | 78 | ||
BC162748.1 | - | 5 | 518 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
