Assay Details
| Gene Symbol: b3galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207101.1 | NP_996984.1 | 2 | 2047 | 113 | |
| GenBank mRNA | BC066477.1 | - | 2 | 2047 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

b3galt2| Gene Symbol: b3galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207101.1 | NP_996984.1 | 2 | 2047 | 113 | |
| GenBank mRNA | BC066477.1 | - | 2 | 2047 | 113 | |
Set Membership: |
|