Assay Details
| Gene Symbol: ppp1r14bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002692.1 | NP_001002692.1 | 2 - 3 | 780 | 132 | |
| GenBank mRNA | BC076513.1 | - | 2 - 3 | 780 | 132 | |
| BC165900.1 | - | 2 - 3 | 318 | 132 | ||
More Information
Set Membership: |
|

ppp1r14bb
Genomic Map