Assay Details
| Gene Symbol: fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003471.1 | NP_001003471.1 | 1 - 2 | 297 | 72 | |
| GenBank mRNA | BC078342.1 | - | 1 - 2 | 297 | 72 | |
| BC165022.1 | - | 1 - 2 | 235 | 72 | ||
More Information
Set Membership: |
|

fkbp14
Genomic Map