Assay Details
| Gene Symbol: cptp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003497.1 | NP_001003497.1 | 1 - 2 | 253 | 131 | |
| GenBank mRNA | BC078407.1 | - | 1 - 2 | 253 | 131 | |
More Information
Set Membership: |
|

cptp
Genomic Map