Assay Details
Gene Symbol: ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003589.2 | NP_001003589.1 | 2 - 3 | 202 | 103 | |
GenBank mRNA | BC078268.1 | - | 2 - 3 | 206 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -




Gene Ontology Categories:
