Assay Details
| Gene Symbol: sgms2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003641.2 | NP_001003641.1 | 2 - 3 | 524 | 75 | |
| GenBank mRNA | BC077132.1 | - | 2 - 3 | 528 | 75 | |
More Information
Set Membership: |
|

sgms2b
Genomic Map