Assay Details
| Gene Symbol: plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003890.3 | NP_001003890.3 | 1 - 2 | 647 | 71 | |
| GenBank mRNA | AY648752.1 | - | 1 - 2 | 606 | 71 | |
| BC098545.1 | - | 1 - 2 | 647 | 71 | ||
| HM210100.1 | - | 1 - 2 | 363 | 71 | ||
More Information
Set Membership: |
|

plk1
Genomic Map