Assay Details
| Gene Symbol: bet1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003998.2 | NP_001003998.1 | 2 - 3 | 345 | 75 | |
| GenBank mRNA | BC080213.1 | - | 2 - 3 | 242 | 75 | |
More Information
Set Membership: |
|

bet1l
Genomic Map