Assay Details
| Gene Symbol: aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004540.1 | NP_001004540.1 | 5 - 6 | 936 | 62 | |
| GenBank mRNA | BC081581.1 | - | 5 - 6 | 936 | 62 | |
| BC152164.1 | - | 5 - 6 | 951 | 62 | ||
| EU155019.1 | - | 5 - 6 | 930 | 62 | ||
More Information
Set Membership: |
|

aldh8a1
Genomic Map