Assay Details
| Gene Symbol: nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004638.1 | NP_001004638.1 | 3 - 4 | 613 | 71 | |
| XM_009302921.3 | XP_009301196.1 | 3 - 4 | 387 | 71 | ||
| GenBank mRNA | BC081382.1 | - | 3 - 4 | 613 | 71 | |
More Information
Set Membership: |
|

nit1
Genomic Map