Assay Details
Gene Symbol: dhrs13a.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004641.1 | NP_001004641.1 | 3 - 4 | 380 | 95 | |
GenBank mRNA | BC081378.1 | - | 3 - 4 | 380 | 95 | |
BC164115.1 | - | 3 - 4 | 400 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
