Assay Details
| Gene Symbol: fkbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004677.2 | NP_001004677.2 | 5 - 6 | 458 | 122 | |
| GenBank mRNA | BC081409.1 | - | 5 - 6 | 414 | 121 | |
| BC155099.1 | - | 5 - 6 | 418 | 122 | ||
| EH538052.1 | - | 5 - 6 | 495 | 122 | ||
| EH560251.1 | - | 1 - 2 | 397 | 122 | ||
More Information
Set Membership: |
|

fkbp2
Genomic Map