Assay Details
Gene Symbol: epha4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005919.2 | NP_001005919.2 | 13 - 14 | 2422 | 67 | |
XM_005171316.4 | XP_005171373.1 | 14 - 15 | 2814 | 67 | ||
XM_005171318.4 | XP_005171375.1 | 13 - 14 | 2705 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
