Assay Details
| Gene Symbol: epha4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005919.2 | NP_001005919.2 | 15 - 16 | 2770 | 87 | |
| NM_001365171.1 | NP_001352100.1 | 15 - 16 | 2797 | 87 | ||
| XM_005171315.4 | XP_005171372.1 | 16 - 17 | 3189 | 87 | ||
| XM_005171316.4 | XP_005171373.1 | 16 - 17 | 3162 | 87 | ||
| XM_005171317.4 | XP_005171374.1 | 15 - 16 | 3079 | 87 | ||
| XM_005171318.4 | XP_005171375.1 | 15 - 16 | 3053 | 87 | ||
More Information
Set Membership: |
|

epha4a
Genomic Map