Assay Details
| Gene Symbol: epha4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005919.2 | NP_001005919.2 | 5 - 6 | 1401 | 66 | |
| NM_001365171.1 | NP_001352100.1 | 5 - 6 | 1401 | 66 | ||
| XM_005171315.4 | XP_005171372.1 | 5 - 6 | 1685 | 66 | ||
| XM_005171316.4 | XP_005171373.1 | 5 - 6 | 1685 | 66 | ||
| XM_005171317.4 | XP_005171374.1 | 5 - 6 | 1683 | 66 | ||
| XM_005171318.4 | XP_005171375.1 | 5 - 6 | 1684 | 66 | ||
| XM_009298712.3 | XP_009296987.1 | 5 - 6 | 1688 | 66 | ||
| GenBank mRNA | BC098529.1 | - | 5 - 6 | 1396 | 66 | |
| KM655611.1 | - | 5 - 6 | 1319 | 66 | ||
More Information
Set Membership: |
|

epha4a
Genomic Map