Assay Details
Gene Symbol: epha4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005919.2 | NP_001005919.2 | 9 - 10 | 1855 | 109 | |
NM_001365171.1 | NP_001352100.1 | 9 - 10 | 1855 | 109 | ||
XM_005171315.4 | XP_005171372.1 | 10 - 11 | 2247 | 109 | ||
XM_005171316.4 | XP_005171373.1 | 10 - 11 | 2247 | 109 | ||
XM_005171317.4 | XP_005171374.1 | 9 - 10 | 2137 | 109 | ||
XM_005171318.4 | XP_005171375.1 | 9 - 10 | 2138 | 109 | ||
XM_009298712.3 | XP_009296987.1 | 9 - 10 | 2142 | 109 | ||
GenBank mRNA | BC098529.1 | - | 9 - 10 | 1850 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
