Assay Details
Gene Symbol: epha4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005919.2 | NP_001005919.2 | 11 - 12 | 2154 | 66 | |
NM_001365171.1 | NP_001352100.1 | 11 - 12 | 2154 | 66 | ||
XM_005171315.4 | XP_005171372.1 | 12 - 13 | 2546 | 66 | ||
XM_005171316.4 | XP_005171373.1 | 12 - 13 | 2546 | 66 | ||
XM_005171317.4 | XP_005171374.1 | 11 - 12 | 2436 | 66 | ||
XM_005171318.4 | XP_005171375.1 | 11 - 12 | 2437 | 66 | ||
XM_009298712.3 | XP_009296987.1 | 11 - 12 | 2441 | 66 | ||
GenBank mRNA | BC098529.1 | - | 11 - 12 | 2149 | 66 | |
U89380.1 | - | 1 - 2 | 197 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
