Assay Details
Gene Symbol: epha4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005919.2 | NP_001005919.2 | 12 - 13 | 2221 | 78 | |
NM_001365171.1 | NP_001352100.1 | 12 - 13 | 2221 | 78 | ||
XM_005171315.4 | XP_005171372.1 | 13 - 14 | 2613 | 78 | ||
XM_005171316.4 | XP_005171373.1 | 13 - 14 | 2613 | 78 | ||
XM_005171317.4 | XP_005171374.1 | 12 - 13 | 2503 | 78 | ||
XM_005171318.4 | XP_005171375.1 | 12 - 13 | 2504 | 78 | ||
XM_009298712.3 | XP_009296987.1 | 12 - 13 | 2508 | 78 | ||
GenBank mRNA | BC098529.1 | - | 12 - 13 | 2216 | 78 | |
U89380.1 | - | 2 - 3 | 264 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
